Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287555.1 | NP_001274484.1 | 1 - 2 | 133 | 86 | |
GenBank mRNA | JO634886.2 | - | 1 - 2 | 341 | 86 | |
JX570673.1 | - | 1 - 2 | 133 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |