Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285764.1 | NP_001272693.1 | 3 - 4 | 500 | 97 | |
XM_005694718.3 | XP_005694775.3 | 3 - 4 | 942 | 97 | ||
GenBank mRNA | HM138664.1 | - | 3 - 4 | 661 | 97 | |
HM138665.1 | - | 3 - 4 | 661 | 97 | ||
HM852511.1 | - | 3 - 4 | 500 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |