Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049650.1 | XP_017905139.1 | 2 - 3 | 717 | 87 | |
GenBank mRNA | JO633672.2 | - | 2 - 3 | 306 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |