Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PQLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039390.1 | XP_017894879.1 | 1 - 2 | 234 | 68 | |
XM_018039391.1 | XP_017894880.1 | 1 - 2 | 234 | 68 | ||
GenBank mRNA | JO391501.2 | - | 2 - 3 | 427 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |