Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063401.1 | XP_017918890.1 | 3 - 4 | 619 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |