Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674919.3 | XP_005674976.1 | 3 - 4 | 617 | 83 | |
GenBank mRNA | JO333044.2 | - | 2 - 3 | 789 | 83 | |
JO578915.1 | - | 2 - 3 | 2108 | 83 | ||
JT705664.1 | - | 2 - 3 | 451 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |