Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064668.1 | XP_017920157.1 | 6 - 7 | 1033 | 72 | |
XM_018064669.1 | XP_017920158.1 | 6 - 7 | 1185 | 72 | ||
XM_018064670.1 | XP_017920159.1 | 6 - 7 | 1030 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |