Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699761.3 | XP_005699818.1 | 3 - 4 | 470 | 99 | |
GenBank mRNA | JO439119.2 | - | 3 - 4 | 468 | 99 | |
JT754263.1 | - | 7 - 8 | 1051 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |