Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC108633183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043754.1 | XP_017899243.1 | 5 - 6 | 607 | 101 | |
XM_018043755.1 | XP_017899244.1 | 526 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |