Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013971438.2 | XP_013826892.2 | 3 - 4 | 270 | 69 | |
XM_018062881.1 | XP_017918370.1 | 3 - 4 | 328 | 69 | ||
GenBank mRNA | KA350368.1 | - | 2 - 3 | 295 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |