Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013977098.2 | XP_013832552.1 | 3 - 4 | 1073 | 100 | |
GenBank mRNA | JO576341.1 | - | 2 - 3 | 598 | 100 | |
KA316325.1 | - | 2 - 3 | 388 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |