Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058037.1 | XP_017913526.1 | 2 - 3 | 244 | 59 | |
XM_018058041.1 | XP_017913530.1 | 2 - 3 | 244 | 59 | ||
XM_018058050.1 | XP_017913539.1 | 2 - 3 | 244 | 59 | ||
XM_018058053.1 | XP_017913542.1 | 2 - 3 | 244 | 59 | ||
GenBank mRNA | JO450443.2 | - | 3 - 4 | 430 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |