Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681903.3 | XP_005681960.3 | 2 - 3 | 354 | 100 | |
XM_005681904.3 | XP_005681961.2 | 2 - 3 | 597 | 100 | ||
XM_005681905.3 | XP_005681962.2 | 2 - 3 | 820 | 100 | ||
XM_005681906.3 | XP_005681963.2 | 2 - 3 | 557 | 100 | ||
GenBank mRNA | JT758801.1 | - | 5 - 6 | 738 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |