Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689730.3 | XP_005689787.1 | 1 - 2 | 656 | 71 | |
GenBank mRNA | JO542256.2 | - | 1 - 2 | 484 | 71 | |
JO584745.2 | - | 2 - 3 | 3566 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |