Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057836.1 | XP_017913325.1 | 2 - 3 | 1193 | 71 | |
GenBank mRNA | KA319296.1 | - | 1 - 2 | 345 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |