Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHISA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013974976.2 | XP_013830430.2 | 2 - 3 | 876 | 82 | |
XM_013974977.2 | XP_013830431.2 | 2 - 3 | 876 | 82 | ||
XM_013974979.2 | XP_013830433.2 | 2 - 3 | 876 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |