Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693629.3 | XP_005693686.2 | 4 - 5 | 647 | 78 | |
XM_013972217.2 | XP_013827671.1 | 5 - 6 | 815 | 78 | ||
XM_013972218.2 | XP_013827672.1 | 4 - 5 | 679 | 78 | ||
GenBank mRNA | JO589125.2 | - | 1 - 2 | 685 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |