Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699520.3 | XP_005699577.1 | 5 - 6 | 673 | 74 | |
XM_018043530.1 | XP_017899019.1 | 4 - 5 | 497 | 74 | ||
GenBank mRNA | JP350874.2 | - | 1 - 2 | 265 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |