Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTNL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050548.1 | XP_017906037.1 | 10 - 11 | 1217 | 55 | |
GenBank mRNA | KA308283.1 | - | 10 - 11 | 1190 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |