Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064327.1 | XP_017919816.1 | 3 - 4 | 560 | 79 | |
GenBank mRNA | JO398689.2 | - | 3 - 4 | 428 | 79 | |
KA315723.1 | - | 2 - 3 | 809 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |