Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676779.3 | XP_005676836.1 | 4 - 5 | 329 | 98 | |
GenBank mRNA | JO582137.1 | - | 4 - 5 | 370 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |