Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679316.3 | XP_005679373.1 | 2 - 3 | 562 | 76 | |
XM_013963385.2 | XP_013818839.1 | 2 - 3 | 479 | 76 | ||
GenBank mRNA | JO369211.2 | - | 1 - 2 | 478 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |