Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680979.3 | XP_005681036.2 | 10 - 11 | 1928 | 86 | |
GenBank mRNA | KA340056.1 | - | 1 - 2 | 408 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |