Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692524.3 | XP_005692581.2 | 1 - 2 | 684 | 103 | |
GenBank mRNA | JO585581.2 | - | 1 - 2 | 380 | 103 | |
JO585582.2 | - | 1 - 2 | 380 | 103 | ||
KA333101.1 | - | 2 - 3 | 1570 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |