Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005694750.3 | XP_005694807.1 | 1 - 2 | 320 | 78 | |
GenBank mRNA | JO642530.2 | - | 1 - 2 | 280 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |