Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICOSLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051877.1 | XP_017907366.1 | 2 - 3 | 218 | 96 | |
XM_018051888.1 | XP_017907377.1 | 2 - 3 | 218 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |