Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPERB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013973131.2 | XP_013828585.2 | 17 - 18 | 1926 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |