Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEPRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674925.3 | XP_005674982.2 | 1 - 2 | 179 | 99 | |
GenBank mRNA | KA323565.1 | - | 1 - 2 | 102 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |