Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055816.1 | XP_017911305.1 | 10 - 11 | 1424 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |