Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C18H19orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692419.3 | XP_005692476.1 | 4 - 5 | 1344 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |