Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055846.1 | XP_017911335.1 | 3 - 4 | 357 | 69 | |
XM_018055847.1 | XP_017911336.1 | 3 - 4 | 359 | 69 | ||
GenBank mRNA | JO400584.2 | - | 5 - 6 | 504 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |