Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOVA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013971571.2 | XP_013827025.1 | 2 - 3 | 303 | 74 | |
GenBank mRNA | JO514999.2 | - | 2 - 3 | 280 | 74 | |
JO631216.2 | - | 716 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |