Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041706.1 | XP_017897195.1 | 4 - 5 | 1377 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |