Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689249.3 | XP_005689306.2 | 3 - 4 | 375 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |