Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047517.1 | XP_017903006.1 | 1 - 2 | 992 | 69 | |
XM_018047518.1 | XP_017903007.1 | 2 - 3 | 716 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |