Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFML2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055754.1 | XP_017911243.1 | 6 - 7 | 1292 | 104 | |
XM_018055755.1 | XP_017911244.1 | 6 - 7 | 1153 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |