Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053511.1 | XP_017909000.1 | 5 - 6 | 880 | 85 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Capra hircus Probe spans exons |