Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCUN1D5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059897.1 | XP_017915386.1 | 5 - 6 | 779 | 102 | |
XM_018059898.1 | XP_017915387.1 | 4 - 5 | 675 | 102 | ||
XM_018059899.1 | XP_017915388.1 | 2 - 3 | 512 | 102 | ||
XM_018059900.1 | XP_017915389.1 | 2 - 3 | 179 | 102 | ||
GenBank mRNA | JO576522.1 | - | 3 - 4 | 467 | 102 | |
JT707780.1 | - | 5 - 6 | 423 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |