Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040766.1 | XP_017896255.1 | 6 - 7 | 610 | 77 | |
XM_018040767.1 | XP_017896256.1 | 6 - 7 | 696 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |