Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698968.3 | XP_005699025.2 | 1 - 2 | 255 | 89 | |
GenBank mRNA | JO578817.1 | - | 1 - 2 | 321 | 89 | |
KA320701.1 | - | 5 - 6 | 1049 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |