Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKZF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013963958.2 | XP_013819412.1 | 5 - 6 | 734 | 58 | |
XM_018048092.1 | XP_017903581.1 | 5 - 6 | 2018 | 58 | ||
XM_018048093.1 | XP_017903582.1 | 6 - 7 | 2112 | 58 | ||
GenBank mRNA | JO606503.2 | - | 2 - 3 | 646 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |