Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFHX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054606.1 | XP_017910095.1 | 6 - 7 | 3307 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |