Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013973561.2 | XP_013829015.2 | 4 - 5 | 524 | 94 | |
GenBank mRNA | JO395476.2 | - | 3 - 4 | 485 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |