Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040863.1 | XP_017896352.1 | 2 - 3 | 676 | 60 | |
XM_018040864.1 | XP_017896353.1 | 2 - 3 | 670 | 60 | ||
XM_018040865.1 | XP_017896354.1 | 3 - 4 | 644 | 60 | ||
GenBank mRNA | JO634469.2 | - | 2 - 3 | 678 | 60 | |
KA306772.1 | - | 3 - 4 | 777 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |