Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048048.1 | XP_017903537.1 | 2 - 3 | 264 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |