Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6H4orf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049278.1 | XP_017904767.1 | 11 - 12 | 1316 | 70 | |
XM_018049279.1 | XP_017904768.1 | 10 - 11 | 1281 | 70 | ||
XM_018049280.1 | XP_017904769.1 | 10 - 11 | 1239 | 70 | ||
XM_018049281.1 | XP_017904770.1 | 10 - 11 | 986 | 70 | ||
XM_018049282.1 | XP_017904771.1 | 10 - 11 | 1116 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |