Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HHLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005688938.2 | XP_005688995.1 | 4 - 5 | 278 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |