Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19H17orf78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693158.3 | XP_005693215.2 | 7 - 8 | 887 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |