Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061710.1 | XP_017917199.1 | 4 - 5 | 534 | 57 | |
XM_018061712.1 | XP_017917201.1 | 4 - 5 | 534 | 57 | ||
XM_018061713.1 | XP_017917202.1 | 4 - 5 | 534 | 57 | ||
GenBank mRNA | JO430744.2 | - | 7 - 8 | 1399 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |