Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPUSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041056.1 | XP_017896545.1 | 3 - 4 | 465 | 76 | |
XM_018041057.1 | XP_017896546.1 | 2 - 3 | 812 | 76 | ||
GenBank mRNA | JO432424.2 | - | 3 - 4 | 494 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |